Learn More
Sino Biological Recombinant Human ICAM-1/CD54 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10346H02H1100
Encompass_Preferred
A DNA sequence encoding the Human ICAM1 (NP_000192.2) (Met1-Glu480) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-240-7900