Learn More
Sino Biological Recombinant Human B7-H2/ICOSLG Protein (hFc Tag), HPLC-verified 1
Supplier:  Sino Biological 11559H02H1
Encompass_Preferred
A DNA sequence encoding the human ICOSLG (O75144-1) (Met1-Ser258) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7106