Learn More
Sino Biological Recombinant Human ICOS Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10344H08H1100
Encompass_Preferred
A DNA sequence encoding the Human ICOS (NP_036224.1) (Met1-Gln134) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8469