Learn More
Sino Biological Anti-B7-H2/ICOSLG Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11559MM01P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICOS Ligand / B7-H2 (rh ICOS Ligand / B7-H2; Catalog#11559-H08H; NP_056074.1; Met1-Ser258) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-7675