Learn More
Sino Biological Recombinant Anti-B7-H2/ICOSLG Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 11559R101F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ICOS Ligand / B7-H2 / ICOSLG (rh ICOS Ligand / B7-H2 / ICOSLG; Catalog#11559-H08H; NP_056074.1; Met1-Ser258) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-0148