Learn More
Sino Biological Human ICOSLG MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11559MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICOS Ligand / B7-H2 / ICOSLG (rh ICOS Ligand / B7-H2 / ICOSLG; Catalog#11559-H08H; NP_056074.1; Met 1-Ser 258). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2791