Learn More
Sino Biological Anti-B7-H2/ICOSLG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310320T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human ICOS ligand fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-8987