Learn More
Sino Biological Recombinant Human B7-H2/ICOSLG Protein (ECD, ICOS ligand), HPLC-verified
Supplier:  Sino Biological 11559HNAH100
Encompass_Preferred
A DNA sequence encoding the human ICOS ligand (NP_056074.1) (Met1-Ser258) was expressed.
Catalog No. 50-237-1358