Learn More
Sino Biological Anti-Insulin-degrading enzyme Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105162T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Insulysin/insulin-degrading enzyme/IDE, and purified by antigen affinity chromatography.
Catalog No. 50-241-1513