Learn More
Proteintech Group Inc Human IDH1 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0006096T
Encompass_Preferred
IDH1, also named as PICD and IDP, belongs to the isocitrate and isopropylmalate dehydrogenases family. IDH1 may be a tumor suppressor that, when mutationally inactivated, contributes to tumorigenesis in part through induction of the HIF-1 pathway(PMID: 19359588). It is a common feature of a major subset of primary human brain cancers. IDH1 mutation is always heterozygotic and IDH1 functions as a dimer, theoretically there will be 25% each wild type and mutant homodimers and 50% heterodimers present in the tumor cells(PMID:21079649).
Catalog No. 50-197-4029