Learn More
ACROBiosystems ACROBiosystems Human IDH1 Protein, His Tag (active enzyme, MALS verified)
Supplier:  ACROBiosystems ID1H51H6100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IDH1, His Tag (ID1-H51H6) is expressed from E.coli cells. It contains AA Met 1 - Leu 414 (Accession # O75874-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.5 kDa. The protein migrates as 45-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6858