Learn More
Biomatik Corporation Human Iduronate-2-Sulfatase (IDS) CLIA Kit
Supplier:  Biomatik Corporation EKU09934
Encompass_Preferred
Human Iduronate-2-Sulfatase (IDS) CLIA Kit for detecting IDS in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P22304. Gene ID: 3423. Target Synonyms: MPS2; SIDS; Hunter Syndrome; Idursulfase; Alpha-L-iduronate sulfate sulfatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8401