Learn More
FUJIFILM Biosciences Human IGF-I (IGF-1), Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Catalog No. 50990744
Encompass_Preferred
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death. Mature human IGF-I is 100% homologous to bovine and porcine IGF-1 proteins.
Catalog No. 50-990-744 Supplier FUJIFILM Biosciences Supplier No. 10034100UG
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death. Mature human IGF-I is 100% homologous to bovine and porcine IGF-1 proteins.