Learn More
Sino Biological Human IFI30 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12623T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFI30 (rh IFI30; Catalog#12623-H08H; P13284; Met1-Lys243). IFI30 specific IgG was purified by Human IFI30 affinity chromatography.
Catalog No. 50-237-1555