Learn More
Mabtech Inc HUMAN IFN-A ELISA PRO KIT
Supplier:  Mabtech Inc 34251HP2
Encompass
ELISAPRO kit for quantification of IFN-a in solution. 2 Plates.CONTENTS: *Biotinylated detection mAbs (MT2/4/6)*Streptavidin-HRP*Recombinant human IFN-α ELISA standard*Substrate (TMB)*Buffers for washing, dilution and standard reconstitution*2 Pre-coated ELISA strip plates (mAbs MT1/3/5)
Catalog No. NC1557206