Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IFN-alpha 2 (alpha2a) Recombinant Protein, PeproTech®

Catalog No. 01672613
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-613 1 mg
01-672-606 100 μg
01-672-628 20 μg
01-672-619 250 μg
01-672-635 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-672-613 Supplier Gibco™ Supplier No. 30002AA1MG
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human IFN-alpha (2a) is a 19.3 kDa protein containing 166 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01563
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 3440
Molecular Weight (g/mol) 19.3 kDa
Name Human IFN-alpha 2 (alpha2a)
Purification Method HPLC, SDS-PAGE
Quantity 1 mg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias alpha-2 A interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1 b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Common Name Interferon alpha-2
Gene Symbol IFNA2
Biological Activity Determined by a cytotoxicity assay using human TF-1 cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MCDLPQTHSL GSRRTLMLLA QMRKISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE
Content And Storage -20°C
Activity Determined by a cytotoxicity assay using human TF-1 cells.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.