Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IFN-alpha A (alpha 2a) Recombinant Protein

Catalog No. PI111001 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
>5x10^6 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI111001 >5x10^6 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI111001 Supplier Invitrogen™ Supplier No. 111001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Store at -70°C or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed. Aliquot and refreeze unused contents to minimize effects of freeze thawing. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of product should be in buffers containing protein such as 0.1% BSA or tissue culture media with serum. Dilution for freezing of material below 2 x 10^5 units/mL is not recommended. One freeze thaw cycle is equivalent to thawing an aliquot prepared from the material received.

Human Interferon-alpha 2a produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 19241 Dalton. The Interferon-a 2a gene was obtained from human leukocytes. The rHuIFN-a 2a is purified by proprietary chromatographic techniques.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01563
Concentration Lot-specific
For Use With (Application) Bioactivity, Immunoprecipitation, Western Blot
Formulation PBS with 0.1% BSA and no preservative
Gene ID (Entrez) 3440
Molecular Weight (g/mol) 19.2 kDa
Name Human IFN-alpha A (alpha 2a)
Purification Method Purified
Quantity >5x10^6 units
Source E. Coli
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias alpha-2a interferon; IFNA; IFNA2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha-2; Interferon alpha-A; interferon, alpha 2; leIF A
Common Name Interferon alpha-2a
Gene Symbol IFNA2
Biological Activity EC50 ̃ 1 U/mL; determined by use of the cytopathic effect inhibition assay described in [Budd et al. (1985) Canc. Chem. Pharm. 12:39 ]. The units are determined by use of a Human IFN Alpha A (Hu IFN- alpha A [2 a]) laboratory standard calibrated to the international reference standard for Human Interferon Alpha A provided by the National Institutes of Health [Meager et al. (2001) J. Immunol Methods. 257(1-2):17-33 ].
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Activity EC50 ~ 1 U/mL; determined by use of the cytopathic effect inhibition assay described in [Budd et al. (1985) Canc. Chem. Pharm. 12:39]. The units are determined by use of a Human IFN Alpha A (Hu IFN- alpha A [2a]) laboratory standard calibrated to the international reference standard for Human Interferon Alpha A provided by the National Institutes of Health [Meager et al. (2001) J. Immunol Methods. 257(1-2):17-33].
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.