Learn More
ACROBiosystems ACROBiosystems Human IFN-alpha 2b (K46R) Protein, Fc Tag (MALS verified)
Catalog No. 502107259
Encompass_Preferred
Recombinant Protein;1MG;Human IFN-alpha 2b (K46R), Fc Tag (IFB-H5253) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 188 (Accession # P01563-1 (K46R)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.7 kDa. The protein migrates as 47-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IFNAR2, His Tag (Cat. No. IF2-H5224) at 5 μg/mL (100 μL/well) can bind Human IFN-alpha 2b (K46R), Fc Tag (Cat. No. IFB-H5253) with a linear range of 2.4-10 ng/mL (QC tested).
Catalog No. 50-210-7259 Supplier ACROBiosystems Supplier No. IFBH52531MG
Recombinant Protein;1MG;Human IFN-alpha 2b (K46R), Fc Tag (IFB-H5253) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 188 (Accession # P01563-1 (K46R)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.7 kDa. The protein migrates as 47-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IFNAR2, His Tag (Cat. No. IF2-H5224) at 5 μg/mL (100 μL/well) can bind Human IFN-alpha 2b (K46R), Fc Tag (Cat. No. IFB-H5253) with a linear range of 2.4-10 ng/mL (QC tested).