Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IFN-alpha A (alpha 2a) (Carrier-Free) Recombinant Protein

Catalog No. PI111011
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI111011 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI111011 Supplier Invitrogen™ Supplier No. 111011
Add to Cart
Edge
Add to Cart

Recombinant Protein

After receipt, the product may be stored at -20°C for short-term use (less than 6 months). For long-term storage, we recommend storing the product at -70°C or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed. Aliquot and refreeze unused contents to minimize effects of freeze thawing. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of product should be in buffers containing protein such as 0.1% BSA or tissue culture media with serum. Dilution for freezing of material below 2 x 10^5 units/mL is not recommended. One freeze thaw cycle is equivalent to thawing an aliquot prepared from the material received.

Human Interferon-alpha 2a produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 19241 Dalton. The Interferon-a 2a gene was obtained from human leukocytes. The rHuIFN-a 2a is purified by proprietary chromatographic techniques.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01563
Concentration 0.2 mg/mL
For Use With (Application) ELISA, Immunoprecipitation, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 3440
Molecular Weight (g/mol) 19.2 kDa
Name Human IFN-alpha A (alpha 2 A) (Carrier-Free)
Quantity 5 μg
Source E. Coli
Endotoxin Concentration <1 EU/ μg
Gene Alias alpha-2 A interferon; IFNA; IFNA2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha-2; Interferon alpha-A; interferon, alpha 2; leIF A
Common Name Interferon alpha-2 A
Gene Symbol IFNA2
Biological Activity EC50 ̃ 1 U/mL; determined by use of the cytopathic effect inhibition assay described in [Rubinstein, S. et al. (1981). J. Virol. 37, 755-758 ]; [Familletti, P.C. et al. (1981). Methods in Enzymology, Vol. 78, 387-394 ]. The units are determined with respect to the international reference standard for human interferon alpha A (Hu-IFN-alpha-A) provided by the National Institutes of Health [see Pestka, S. (1986). Methods in Enzymology, Academic Press, New York 119, 14-23 ]. Unit of activity measured on bovine MDBK cells with vesicular stomatitis virus (VSV).
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.