Learn More
ACROBiosystems Human IFN-alpha / beta R1, His Tag (IF1-H5225) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Lys 436 (Accession # P17181-1).
Catalog No. 502230684
Encompass_Preferred
Interferon alpha/beta receptor 1 ( IFNAR1) is also known as Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1 (CRF2-1), Type I interferon receptor 1, IFNAR, which belongs to the type II cytokine receptor family. IFNAR1 /IFNAR contains four fibronectin type-III domains. IFNAR1 associates with IFNAR2 to form the type I interferon receptor. IFNAR1 is receptor for interferons alpha and beta. IFNAR1 can transduce IFNB signals without the help of IFNAR2, and not activating the Jak-STAT pathway.
This item is not returnable. View return policy
Catalog No. 50-223-0684 Supplier ACROBiosystems Supplier No. IF1H52251MG
Interferon alpha/beta receptor 1 ( IFNAR1) is also known as Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1 (CRF2-1), Type I interferon receptor 1, IFNAR, which belongs to the type II cytokine receptor family. IFNAR1 /IFNAR contains four fibronectin type-III domains. IFNAR1 associates with IFNAR2 to form the type I interferon receptor. IFNAR1 is receptor for interferons alpha and beta. IFNAR1 can transduce IFNB signals without the help of IFNAR2, and not activating the Jak-STAT pathway.