Learn More
Raybiotech Inc Human IFN-alpha / beta R2 ELISA Kit
Supplier:  Raybiotech Inc ELHIFNABR21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.16 ng/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM9, CD244, APRIL, BMP-2, BMP-9, Cathepsin L, CD200, CD97, Chemerin, C5a, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGFR, FABP2, FAP, FGF-19, IFG-2R, IL-1R6, IL-24, IL-33, Kallikrein 14, Legumain, LOX-1, MBL, Neprilysin, Notch-1, NOV, Osteoactivin, PG-1, PGRPs, SerpinA4, sFRP-3, Thrombomodulin, TLR2, Transferrin, WIF-1
Catalog No. 50-194-9372