Learn More
ACROBiosystems ACROBiosystems Human IFN-alpha/beta R2 Protein, His Tag
Supplier:  ACROBiosystems IF2H5224100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IFNAR2, His Tag (IF2-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Lys 243 (Accession # P48551-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 34-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6659