Learn More
Raybiotech Inc Human IFN alpha ELISA Kit
Supplier:  Raybiotech Inc ELHIFNA1
Encompass_Preferred
For cell culture supernatants, Heparin and/or EDTA treated plasma, and serum samples. Citrate is not recommended. Sensitivity: 0.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human IFN-alpha. Other species not determined.
Catalog No. 50-194-9371