Learn More
PBL Assay Science VeriKine Human Interferon Alpha (IFN-Alpha, IFNa) ELISA, Tissue Culture Media (TCM) [1x96-well plate]
Supplier:  PBL Assay Science 411001
Encompass_Preferred
VeriKine Human Interferon Alpha ELISA Kit is manufactured by PBL Assay Science (Catalog #41100-1) for research use only (RUO) and packaged as a 1x96-well plate. This IFNa ELISA is Tissue Culture Media (TCM) compatible, quantifies Hu IFN-Alpha, and has an assay range of 12.5-500 pg/mL or 156-5,000 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
Catalog No. 50-153-8054