Learn More
PBL Assay Science Human Interferon Alpha (IFN-Alpha, IFNa) Sampler Set [1e5 (100,000) units/vial]
Catalog No. 501537945 Shop All PBL Assay Science Products
Encompass_Preferred
Recombinant Human Interferon Alpha proteins are manufactured by PBL Assay Science (Catalog #11002-1) for research use only (RUO) and packaged in a sampler set. This protein set contains 12 distinct human IFN-Alpha subtypes. cDNAs of IFNs are obtained from human leukocyte mRNA, expressed in E.coli and purified. These cytokines are packaged in 1e5 (100,000) units/vial.
Catalog No. 50-153-7945 Supplier PBL Assay Science Supplier No. 110021
Recombinant Human Interferon Alpha proteins are manufactured by PBL Assay Science (Catalog #11002-1) for research use only (RUO) and packaged in a sampler set. This protein set contains 12 distinct human IFN-Alpha subtypes. cDNAs of IFNs are obtained from human leukocyte mRNA, expressed in E.coli and purified. These cytokines are packaged in 1e5 (100,000) units/vial.