Learn More
R&D Systems™ Human IFN-beta ELISA Kit



Description
Includes
- Pre-coated microtiter plate(s)
- Plate sealers
- Wash Solution Concentrate
- Human Interferon Alpha Standard, 10,000 pg/ml
- Dilution Buffer
- Antibody Concentrate
- HRP Conjugate Concentrate
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Assay Sensitivity | 25pg/mL |
| Conjugate | Biotin |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Line | R&D Systems |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant |
| Specificity | Human IFN-beta |
| Sufficient For | 5 x 96-well Plates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.