Learn More
PBL Assay Science VeriKine Human Inteferon Beta (IFN-Beta, IFNb), Tissue Culture Media (TCM) (TCM) [1x96 well plate]
Supplier:  PBL Assay Science 414101
Encompass_Preferred
VeriKine Human Interferon Beta ELISA Kit is manufactured by PBL Assay Science (Catalog #41410-1) for research use only (RUO) and packaged as a 1x96-well plate. This IFNb ELISA is Tissue Culture Media (TCM) compatible, quantifies Hu IFN-Beta, and has an assay range of 50-4000 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 8%.
Catalog No. 50-153-8062