Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IFN-beta Quantikine ELISA Kit
SDP

Catalog No. DIFNB0
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DIFNB0 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DIFNB0 Supplier R&D Systems™ Supplier No. DIFNB0

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Human IFN-beta Quantikine ELISA Kit is designed to measure human IFN-beta in cell culture supernates.

The Human IFN-beta Quantikine ELISA Kit quantifies human IFN-beta in cell culture supernates (50 ul).

Specifications

Assay Range 7.81 - 500pg/mL
Assay Sensitivity 0.781pg/mL
Sample Type Cell Culture Supernatant
Specificity Natural and Recombinant Human IFN-beta
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3456
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.