Learn More
PBL Assay Science VeriKine-HS Human Interferon Beta (IFN-Beta, IFNb) ELISA, Plasma/Serum/Tissue Culture Media (TCM) (High Sensitivity) [1x96-well plate]
Supplier:  PBL Assay Science 414151
Encompass
VeriKine-HS Human Interferon Beta Serum ELISA Kit is manufactured by PBL Assay Science (Catalog #41415-1) for research use only (RUO) and packaged as a 1x96-well plate. This IFNb ELISA is Plasma, Serum, Autoimmune Sera, and Tissue Culture Media (TCM) compatible, quantifies Hu IFN-Beta, and has an assay range of 1.2-150 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10% and is an externally validated assay.
Catalog No. 50-153-8064