Learn More
CELLULAR TECHNOLOGY LTD Human IFN-γ BCIP Single-Color ELISPOT
SDP
Supplier:  CELLULAR TECHNOLOGY LTD HIFNGP1M/2
Encompass
This optimized ELISPOT assay reliably detects and measures human and some NHP IFN-γ secretions by stimulated effector cells in frequencies as low as one in a million.
Catalog No. NC1787696