Learn More
Abcam Human IFN gamma ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB460251X96TESTS
Encompass_Preferred
Human IFN gamma ELISA Kit ab46025 is used to quantify Human IFN gamma with a sensitivity of 0.59 ng/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2251289