Learn More
ACROBiosystems Human IFN-Gamma (IFNG) Protein, Premium Grade, HEK293-Expressed, 20 μg
Supplier: ACROBiosystems IFGH421120UG
Recombinant human interferon-gamma (IFN-γ) premium-grade protein produced in HEK293 cells. It contains amino acids Gln 24-Gln 166, is supplied lyophilized from PBS with trehalose, and is intended for cell culture and early-stage discovery or preclinical manufacturing workflows. The product is quality-tested for sterility and endotoxin.
- Expressed in human HEK293 cells for human-like post-translational modifications.
- Contains amino acids Gln 24-Gln 166 (Accession: P01579).
- Purity greater than 95% by SDS-PAGE and greater than 90% by SEC-MALS.
- Lyophilized formulation in PBS with trehalose, 0.22 μm filtered.
- Store lyophilized at -20°C or lower; reconstituted material stable at -70°C for up to 3 months.
- Suitable for cell culture applications and early-stage discovery or preclinical manufacturing workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.