Learn More
Sino Biological Recombinant Human IFN gamma Protein, HPLC-verified
Supplier:  Sino Biological 11725HNAE20
Encompass_Preferred
A DNA sequence encoding the human IFNG (NP_000610.2) (Gln24-Gln166) was expressed with an N-terminal Met.
Catalog No. 50-228-2149