Learn More
Raybiotech Inc Human IFN-gamma R1 ELISA Kit
Supplier:  Raybiotech Inc ELHIFNGR11
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 24 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ALK-1, B7-H2, BLAME, BMP-8, CD28, Common beta Chain (CD131), Contactin-1, Desmoglein-1, Desmoglein-3, EDAR, EphA1, EphB6, Ephrin-B3, Epiregulin, FGF-12, FGF-17, FOLR2, Galectin-8, Glypican 1, Glypican 5, GHR, IL-22 R alpha 1, IL-22BP, IL-23 R, IL-31 RA, IL-7R alpha, Integrin alpha 5, MDM2, Nectin-1, NKp30, Nogo Receptor, Notch-3, OSM R beta, Prolactin R, RELT, Ryk, Semaphorin 6D, Semaphorin 7A, Siglec-11.
Catalog No. 50-194-9375