Learn More
ACROBiosystems ACROBiosystems Human IFN-gamma R1 / IFNGR1 Protein, Fc Tag
Catalog No. 502106658
Encompass_Preferred
Recombinant Protein;200UG;Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.4 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human IFN-gamma, Tag Free (Cat. No. IFG-H4211) at 5 μg/mL (100 μL/well) can bind Human IFN-gamma R1, Fc Tag (Cat. No. IF1-H5254) with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-210-6658 Supplier ACROBiosystems Supplier No. IF1H5254200UG
Recombinant Protein;200UG;Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.4 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human IFN-gamma, Tag Free (Cat. No. IFG-H4211) at 5 μg/mL (100 μL/well) can bind Human IFN-gamma R1, Fc Tag (Cat. No. IF1-H5254) with a linear range of 1-10 ng/mL (QC tested).