Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IFN-omega Recombinant Protein

Catalog No. PI113951
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI113951 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI113951 Supplier Invitrogen™ Supplier No. 113951
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Store at -20°C or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed. Aliquot and refreeze unused contents to minimize effects of freeze thawing. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of product should be in buffers containing protein such as 0.1% BSA or tissue culture media with serum.

IFN-omega is a type I interferon that can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-alpha, IFN-beta, and IFN-omega, signal through the IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities. IFN-omega exhibits about 75% sequence homology with IFN-alpha, and contains two conserved disulfide bonds that are necessary for full biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05000
Concentration Lot-specific
For Use With (Application) ELISA, Immunoprecipitation, Western Blot
Formulation PBS with 2% BSA and no preservative
Gene ID (Entrez) 3467
Molecular Weight (g/mol) 20 kDa
Name Human IFN-omega
Quantity 25 μg
Source E. Coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias IFN o; IFN ?; IFNo; IFNomega; IFN-omega 1, interferon omega-1; IFNW; IFNW1; IFN?; Interferon; Interferon alpha-II-1; Interferon o; Interferon OMEGA; interferon omega 1; interferon omega-1; Interferon ?; interferon, omega 1
Common Name IFN omega
Gene Symbol IFNW1
Biological Activity EC50 ̃ 1 U/mL; determined by use of the cytopathic effect inhibition assay described in [Rubinstein, S. et al. (1981). J. Virol. 37, 755-758 ]; [Familletti, P.C. et al. (1981). Methods in Enzymology, Vol. 78, 387-394 ]. The units are determined with respect to the international reference standard for human interferon alpha A (Hu-IFN-alpha-A) provided by the National Institutes of Health [see Pestka, S. (1986). Methods in Enzymology, Academic Press, New York 119, 14-23 ]. Unit of activity measured on bovine MDBK cells with vesicular stomatitis virus (VSV).SV).
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.