Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IFN-omega Recombinant Protein, PeproTech®

Catalog No. 30002J100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30002J100UG 100 μg
300-02J-1MG 1 mg
30002J20UG 20 μg
30002J250UG 250 μg
30002J500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 30002J100UG Supplier Gibco™ Supplier No. 30002J100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IFN-omega is a 19.9 kDa protein consisting of 172 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IFN-omega is a type I interferon that can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-alpha, IFN-beta, and IFN-omega, signal through the IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities. IFN-omega exhibits about 75% sequence homology with IFN-alpha, and contains two conserved disulfide bonds that are necessary for full biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05000
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3467
Molecular Weight (g/mol) 19.9 kDa
Name Human IFN-omega
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias IFN o; IFN ω; IFNo; IFNomega; IFN-omega 1, interferon omega-1; IFNW; IFNW1; IFNω; Interferon; Interferon alpha-II-1; Interferon o; Interferon OMEGA; interferon omega 1; interferon omega-1; Interferon ω; interferon, omega 1
Common Name IFN omega
Gene Symbol IFNW1
Biological Activity The ED50 was determined by a cytotoxicity assay using Human TF-1 cells is ≤ 0.01 ng/ml, corresponding to a specific activity of ≥ 1 x 10^8 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence CDLPQNHGLL SRNTLVLLHQ MRRISPFLCL KDRRDFRFPQ EMVKGSQLQK AHVMSVLHEM LQQIFSLFHT ERSSAAWNMT LLDQLHTGLH QQLQHLETCL LQVVGEGESA GAISSPALTL RRYFQGIRVY LKEKKYSDCA WEVVRMEIMK SLFLSTNMQE RLRSKDRDLG SS
Content And Storage -20°C
Activity The ED50 was determined by a cytotoxicity assay using Human TF-1 cells is ≤ 0.01 ng/ml, corresponding to a specific activity of ≥ 1 x 10^8 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.