Learn More
Mabtech Inc HUMAN IFN-Y BIOTIN
Supplier:  Mabtech Inc 34206250
Encompass
Monoclonal antibody specific for IFN-γ This monoclonal antibody enables sensitive and specific detection of human IFN-γ in immunoassays such as ELISA and ELISpot. We recommend 1-D1K as capture antibody in ELISA and ELISpot in combination with biotinylated detection antibody 7-B6-1
Catalog No. NC9865659