Learn More
PBL Assay Science VeriKine Human Interferon Alpha (IFN-Alpha, IFNa) ELISA, Multi-Subtype, Plasma/Serum/Tissue Culture Media (TCM) [1x96-well plate]
Supplier:  PBL Assay Science 411101
Encompass_Preferred
VeriKine Human Interferon Alpha Multi-Subtype Serum ELISA Kit is manufactured by PBL Assay Science (Catalog #41110-1) for research use only (RUO) and packaged as a 1x96-well plate. This IFNa Multi-Subtype Serum ELISA is Serum, Plasma, and Tissue Culture Media (TCM) compatible, quantifies 11 of 12 human IFN-Alpha subtypes, and has an assay range of 12.5-500 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
Catalog No. 50-153-8058