Learn More
Sino Biological Recombinant Anti-Interferon alpha 2/IFNA2 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 13833R001H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Interferon alpha 2/IFNA2 (rh Interferon alpha 2/IFNA2; Catalog#13833-HNAY; P01563.1; Cys24-Glu188) and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1632