Learn More
Sino Biological Human IFNA2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13833RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNA2 (rh IFNA2; Catalog#13833-HNAY; P01563.1; Cys24-Glu188). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3477