Learn More
Sino Biological Recombinant Anti-interferon-alpha/IFNA5 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10342R10220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFNA5/IFNaG (rh IFNA5/IFNaG; Catalog#10342-H08H; P01569; Met1-Glu189).
Catalog No. 50-240-8694