Learn More
Sino Biological Anti-Interferon alpha/beta receptor 1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13222MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFNAR1 (rh IFNAR1; Catalog#13222-H08H; P17181-1; Met1-Lys436). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4276