Learn More
Sino Biological Anti-Interferon alpha/beta receptor 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13222T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNAR1 (rh IFNAR1; Catalog#13222-H08H; P17181-1; Met1-Lys436). IFNAR1 specific IgG was purified by Human IFNAR1 affinity chromatography.
Catalog No. 50-255-4273