Learn More
Sino Biological Anti-IFN-beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10704RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNB1 / IFN-beta / Interferon beta (rh IFNB1 / IFN-beta / Interferon beta; Catalog#10704-HNAS; P01574; Met1-Asn187). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1581