Learn More
Sino Biological Human IFNG ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502373813
Encompass_Preferred
Each vial contains 19 ng of recombinant Human Interferon Gamma/IFN gamma/IFNG. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial in Dilution Buffer, and a high standard of 1400 pg/mL is recommended.
Catalog No. 50-237-3813 Supplier Sino Biological Supplier No. SEKA117255
Each vial contains 19 ng of recombinant Human Interferon Gamma/IFN gamma/IFNG. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial in Dilution Buffer, and a high standard of 1400 pg/mL is recommended.