Learn More
Sino Biological Recombinant Anti-IFN gamma Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 11725R003C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Interferon Gamma/IFN gamma/IFNG (rh Interferon Gamma/IFN gamma/IFNG; Catalog#11725-HNAS; NP_000610.2; Met1-Gln166) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-1228