Learn More
Sino Biological Human IFNG MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11725R23820
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Interferon Gamma/IFN gamma/IFNG (rh Interferon Gamma/IFN gamma/IFNG; Catalog#11725-HNAS; NP_000610.2; Met1-Gln166).
Catalog No. 50-237-2675