Learn More
Sino Biological Anti-IFN gamma Antibody (Biotin), Rabbit Polyclonal
Supplier:  Sino Biological 11725RP02B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNG (rh IFNG; Catalog#11725-HNAS; NP_000610.2; Met 1-Gln 166). IFNG IgG was purified by human IFNG affinity chromatography, and then biotinylated.
Catalog No. 50-255-4598