Learn More
Sino Biological Human IFNG PAb 100ug (Sino Biological)
Supplier:  Sino Biological 11725RP02H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNG (rh IFNG; Catalog#11725-HNAS; NP_000610.2; Met 1-Gln 166). IFNG IgG was purified by human IFNG affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-237-1067